Gene Transcripts
- Gene Symbol
- EIF4A2
- Entrez Gene ID
- 1974
- Gene Name
- eukaryotic translation initiation factor 4A2
- Gene Aliases
- BM-010, DDX2B, EIF4A, EIF4F, eIF-4A-II, eIF4A-II
- Chromosome Location
- Chr. 3: 186783572 - 186789897 on Build GRCh38
- UniGene ID
- Hs.518475
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC039344.1 |
- |
1 |
1395 |
|
View Full Product Details