Gene Transcripts
- Gene Symbol
- THAP5
- Entrez Gene ID
- 168451
- Gene Name
- THAP domain containing 5
- Gene Aliases
- Chromosome Location
- Chr. 7: 108562144 - 108569768 on Build GRCh38
- UniGene ID
- Hs.707689
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC016973.1 |
- |
|
221 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BE889151.1 |
- |
|
200 |
|
Chr. - - [-] |
View Full Product Details