Gene Transcripts

Gene Symbol
THAP5
Entrez Gene ID
168451
Gene Name
THAP domain containing 5
Gene Aliases
Chromosome Location
Chr. 7: 108562144 - 108569768 on Build GRCh38
UniGene ID
Hs.707689
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA BC016973.1 - 256


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BE889151.1 - 235 Chr. - - [-]

View Full Product Details