Gene Transcripts
- Gene Symbol
- RHOF
- Entrez Gene ID
- 54509
- Gene Name
- ras homolog family member F, filopodia associated
- Gene Aliases
- ARHF, RIF
- Chromosome Location
- Chr. 12: 121777754 - 121793688 on Build GRCh38
- UniGene ID
- Hs.707579
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK130459.1 |
- |
|
673 |
|
View Full Product Details