Gene Transcripts

Gene Symbol
RAB28
Entrez Gene ID
9364
Gene Name
RAB28, member RAS oncogene family
Gene Aliases
CORD18
Chromosome Location
Chr. 4: 13293479 - 13484365 on Build GRCh38
UniGene ID
Hs.656060
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005248218.4 XP_005248275.1 5 3845


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AK001995.1 - 1 1863 Chr. 4 - 13456451 - 13458546 [-]

View Full Product Details