Gene Transcripts
- Gene Symbol
- PCP2
- Entrez Gene ID
- 126006
- Gene Name
- Purkinje cell protein 2
- Gene Aliases
- GPSM4
- Chromosome Location
- Chr. 19: 7631614 - 7636967 on Build GRCh38
- UniGene ID
- Hs.591400
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055767.1 |
- |
1 |
849 |
|
View Full Product Details