Gene Transcripts
- Gene Symbol
- ADRA1B
- Entrez Gene ID
- 147
- Gene Name
- adrenoceptor alpha 1B
- Gene Aliases
- ADRA1, ALPHA1BAR
- Chromosome Location
- Chr. 5: 159916733 - 159989205 on Build GRCh38
- UniGene ID
- Hs.368632
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648936.1 |
- |
1 |
2609 |
|
View Full Product Details