Gene Transcripts
- Gene Symbol
- RASGRF1
- Entrez Gene ID
- 5923
- Gene Name
- Ras protein specific guanine nucleotide releasing factor 1
- Gene Aliases
- CDC25, CDC25L, GNRP, GRF1, GRF55, H-GRF55, PP13187, ras-GRF1
- Chromosome Location
- Chr. 15: 78959947 - 79090873 on Build GRCh38
- UniGene ID
- Hs.459035
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL359931.1 |
- |
1 |
1333 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA119282.1 |
- |
1 |
54 |
|
Chr. 15 - 78979837 - 78980387 [-] |
View Full Product Details