Gene Transcripts

Gene Symbol
SYTL2
Entrez Gene ID
54843
Gene Name
synaptotagmin like 2
Gene Aliases
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location
Chr. 11: 85694221 - 85811182 on Build GRCh38
UniGene ID
Hs.369520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK056722.1 - 1 1317


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AL700484.1 - 1 423 Chr. 11 - 85705220 - 85705872 [-]

View Full Product Details