Gene Transcripts
- Gene Symbol
- TECR
- Entrez Gene ID
- 9524
- Gene Name
- trans-2,3-enoyl-CoA reductase
- Gene Aliases
- GPSN2, MRT14, SC2, TER
- Chromosome Location
- Chr. 19: 14527760 - 14565980 on Build GRCh38
- UniGene ID
- Hs.515642
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC034484.1 |
- |
1 |
2156 |
|
View Full Product Details