Gene Transcripts

Gene Symbol
TRIM36
Entrez Gene ID
55521
Gene Name
tripartite motif containing 36
Gene Aliases
HAPRIN, RBCC728, RNF98
Chromosome Location
Chr. 5: 115124762 - 115180546 on Build GRCh38
UniGene ID
Hs.519514
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK125426.1 - 2 2845

View Full Product Details