Gene Transcripts
- Gene Symbol
- C16orf72
- Entrez Gene ID
- 29035
- Gene Name
- chromosome 16 open reading frame 72
- Gene Aliases
- PRO0149
- Chromosome Location
- Chr. 16: 9091329 - 9119698 on Build GRCh38
- UniGene ID
- Hs.221497
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM055234.1 |
- |
1 |
338 |
|
Chr. 16 - 9112754 - 9113180 [+] |
GenBankmRNA |
DB571277.1 |
- |
1 |
386 |
|
Chr. 16 - 9112704 - 9113180 [+] |
GenBankmRNA |
BM127137.1 |
- |
|
178 |
|
Chr. - - [-] |
GenBankmRNA |
CD677786.1 |
- |
1 |
508 |
|
Chr. 16 - 9112582 - 9113132 [+] |
View Full Product Details