Gene Transcripts

Gene Symbol
CUX1
Entrez Gene ID
1523
Gene Name
cut like homeobox 1
Gene Aliases
CASP, CDP, CDP/Cut, CDP1, COY1, CUTL1, CUX, Clox, Cux/CDP, GOLIM6, Nbla10317, p100, p110, p200, p75
Chromosome Location
Chr. 7: 101815904 - 102283958 on Build GRCh38
UniGene ID
Hs.191482
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001202543.1 NP_001189472.1 24 10643
NM_181552.3 NP_853530.2 24 10521
XM_005250150.2 XP_005250207.1 24 11039
XM_006715854.2 XP_006715917.1 23 10976
XM_011515823.2 XP_011514125.1 24 10734
XM_011515825.1 XP_011514127.1 24 10302
XM_017011760.1 XP_016867249.1 23 10536
GenBank mRNA AK056307.1 - 1 2028

View Full Product Details