Gene Transcripts
- Gene Symbol
- KHK
- Entrez Gene ID
- 3795
- Gene Name
- ketohexokinase
- Gene Aliases
- Chromosome Location
- Chr. 2: 27086738 - 27100751 on Build GRCh38
- UniGene ID
- Hs.567297
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AB209518.1 |
- |
1 |
832 |
|
View Full Product Details