Gene Transcripts
- Gene Symbol
- ZDHHC1
- Entrez Gene ID
- 29800
- Gene Name
- zinc finger DHHC-type containing 1
- Gene Aliases
- C16orf1, HSU90653, ZNF377
- Chromosome Location
- Chr. 16: 67394419 - 67416494 on Build GRCh38
- UniGene ID
- Hs.624541
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC045169.2 |
- |
2 |
1070 |
|
View Full Product Details