Gene Transcripts

A Gene Symbol is not available because the non-coding siRNA was designed to a GenBank RNA or EST accession that does not map to an Entrez Gene ID. Non-coding siRNAs are designed to RefSeq (NR_) or to curated sequences from RNAdb represented by GenBank RNA or ESTdb accessions.


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA AF071167.3 - 1 335 Chr. 7 - 27188112 - 27189293 [+]
GenBankmRNA BI045261.1 - 1 164 Chr. 7 - 27188287 - 27188833 [+]
GenBankmRNA AF071170.3 - 1 373 Chr. 7 - 27188074 - 27189293 [+]

View Full Product Details