Gene Transcripts
- Gene Symbol
- G3BP1
- Entrez Gene ID
- 10146
- Gene Name
- G3BP stress granule assembly factor 1
- Gene Aliases
- G3BP, HDH-VIII
- Chromosome Location
- Chr. 5: 151771915 - 151811352 on Build GRCh38
- UniGene ID
- Hs.3353
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK091592.1 |
- |
2 |
2312 |
|
View Full Product Details