Gene Transcripts
- Gene Symbol
- LYST
- Entrez Gene ID
- 1130
- Gene Name
- lysosomal trafficking regulator
- Gene Aliases
- CHS, CHS1
- Chromosome Location
- Chr. 1: 235661031 - 235883708 on Build GRCh38
- UniGene ID
- Hs.532411
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_102436.2 |
|
4 |
2861 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR624421.1 |
- |
1 |
798 |
|
Chr. 1 - 235826343 - 235827784 [-] |
View Full Product Details