Gene Transcripts

Gene Symbol
KANSL1L
Entrez Gene ID
151050
Gene Name
KAT8 regulatory NSL complex subunit 1 like
Gene Aliases
C2orf67, MSL1v2
Chromosome Location
Chr. 2: 210021421 - 210171370 on Build GRCh38
UniGene ID
Hs.282260
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_005246334.4 XP_005246391.1 6 2607
XM_006712320.3 XP_006712383.1 6 2689


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BX376230.2 - 1 125 Chr. 2 - 210096310 - 210097005 [-]
GenBankmRNA CR616459.1 - 1 125 Chr. 2 - 210096310 - 210097005 [-]

View Full Product Details