Gene Transcripts
- Gene Symbol
- GEN1
- Entrez Gene ID
- 348654
- Gene Name
- GEN1, Holliday junction 5' flap endonuclease
- Gene Aliases
- Gen
- Chromosome Location
- Chr. 2: 17753687 - 17785920 on Build GRCh38
- UniGene ID
- Hs.467793
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK093631.1 |
- |
1 |
551 |
|
BC038243.1 |
- |
1 |
815 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
CR617603.1 |
- |
1 |
1414 |
|
Chr. 2 - 17753296 - 17755047 [+] |
GenBankmRNA |
DR000919.1 |
- |
1 |
543 |
|
Chr. 2 - 17754174 - 17754873 [+] |
GenBankmRNA |
BF979651.2 |
- |
1 |
553 |
|
Chr. 2 - 17754166 - 17754846 [+] |
GenBankmRNA |
DB104754.1 |
- |
1 |
551 |
|
Chr. 2 - 17754160 - 17754731 [+] |
View Full Product Details