Gene Transcripts

Gene Symbol
EPHA6
Entrez Gene ID
285220
Gene Name
EPH receptor A6
Gene Aliases
EHK-2, EHK2, EK12, EPA6, HEK12, PRO57066
Chromosome Location
Chr. 3: 96814581 - 97761532 on Build GRCh38
UniGene ID
Hs.272208
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006713592.3 XP_006713655.1 19 15837
XM_017006210.1 XP_016861699.1 18 15711
XM_017006211.1 XP_016861700.1 20 15862
XM_017006212.1 XP_016861701.1 19 15531
XM_017006213.1 XP_016861702.1 19 15889
XM_017006214.1 XP_016861703.1 18 15157
XM_017006215.1 XP_016861704.1 14 14186
XM_017006216.1 XP_016861705.1 13 14136
XM_017006217.1 XP_016861706.1 15 14233
XM_017006218.1 XP_016861707.1 13 14036
XM_017006220.1 XP_016861709.1 15 14201
XM_017006221.1 XP_016861710.1 13 14147
XM_017006222.1 XP_016861711.1 14 14195
GenBank mRNA AK000957.1 - 1 1025

View Full Product Details