Gene Transcripts
- Gene Symbol
- AKTIP
- Entrez Gene ID
- 64400
- Gene Name
- AKT interacting protein
- Gene Aliases
- FT1, FTS
- Chromosome Location
- Chr. 16: 53491028 - 53504411 on Build GRCh38
- UniGene ID
- Hs.380897
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK098222.1 |
- |
1 |
167 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DB250764.1 |
- |
1 |
167 |
|
Chr. 16 - 53493397 - 53493968 [-] |
GenBankmRNA |
DA523849.1 |
- |
1 |
425 |
|
Chr. 16 - 53493651 - 53494225 [-] |
View Full Product Details