Gene Transcripts
- Gene Symbol
- TMEM43
- Entrez Gene ID
- 79188
- Gene Name
- transmembrane protein 43
- Gene Aliases
- ARVC5, ARVD5, EDMD7, LUMA
- Chromosome Location
- Chr. 3: 14124940 - 14143680 on Build GRCh38
- UniGene ID
- Hs.517817
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK026869.1 |
- |
1 |
202 |
|
View Full Product Details