Gene Transcripts

Gene Symbol
WDFY2
Entrez Gene ID
115825
Gene Name
WD repeat and FYVE domain containing 2
Gene Aliases
PROF, WDF2, ZFYVE22
Chromosome Location
Chr. 13: 51584194 - 51766799 on Build GRCh38
UniGene ID
Hs.208550
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_011534914.1 XP_011533216.1 12 3341
XM_011534915.2 XP_011533217.1 11 6149
XM_017020386.1 XP_016875875.1 12 11873
GenBank mRNA AK095358.1 - 1 456

View Full Product Details