Gene Transcripts

Gene Symbol
C12orf40
Entrez Gene ID
283461
Gene Name
chromosome 12 open reading frame 40
Gene Aliases
HEL-206, HEL-S-94
Chromosome Location
Chr. 12: 39626167 - 39908300 on Build GRCh38
UniGene ID
Hs.648205
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_135051.1 14 1967
GenBank mRNA AK097445.1 - 14 1916
EU668334.1 - 14 1916
GQ472216.1 - 14 1916

View Full Product Details