Gene Transcripts
- Gene Symbol
- IRS4
- Entrez Gene ID
- 8471
- Gene Name
- insulin receptor substrate 4
- Gene Aliases
- IRS-4, PY160
- Chromosome Location
- Chr. X: 108719949 - 108736584 on Build GRCh38
- UniGene ID
- Hs.407141
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL832136.1 |
- |
1 |
4001 |
|
View Full Product Details