Gene Transcripts

Gene Symbol
SNHG5
Entrez Gene ID
387066
Gene Name
small nucleolar RNA host gene 5
Gene Aliases
C6orf160, LINC00044, NCRNA00044, U50HG, bA33E24.2
Chromosome Location
Chr. 6: 85677007 - 85678733 on Build GRCh38
UniGene ID
Hs.292457
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK311192.1 - 1 502
BC009220.2 - 1 471
BC108291.1 - 1 489


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA BG422538.1 - 1 438 Chr. 6 - 85677824 - 85678668 [-]
GenBankmRNA AL525144.3 - 1 494 Chr. 6 - 85677103 - 85678710 [-]
GenBankmRNA BM728819.1 - 1 490 Chr. 6 - 85677811 - 85678721 [-]
GenBankmRNA BM702833.1 - 1 375 Chr. 6 - 85677474 - 85678606 [-]
GenBankmRNA CD639840.1 - 170 Chr. - - [-]
GenBankmRNA AB017710.3 - 1 1129 Chr. 6 - 85677007 - 85679361 [-]
GenBankmRNA HX785786.1 - 2 349 Chr. 6 - 85677147 - 85678602 [-]
GenBankmRNA BG107883.1 - 1 31 Chr. 6 - 85677082 - 85678262 [-]
GenBankmRNA BU944864.1 - 1 353 Chr. 6 - 85677082 - 85678584 [-]
GenBankmRNA CD676465.1 - 1 40 Chr. 6 - 85677082 - 85678252 [-]
GenBankmRNA HX789156.1 - 2 349 Chr. 6 - 85677147 - 85678602 [-]
GenBankmRNA BE931917.1 - 1 138 Chr. 6 - 85677155 - 85678356 [-]
GenBankmRNA BG697433.1 - 1 489 Chr. 6 - 85677426 - 85678720 [-]
GenBankmRNA BG472221.1 - 490 Chr. - - [-]

View Full Product Details