Gene Transcripts
- Gene Symbol
- CBY1
- Entrez Gene ID
- 25776
- Gene Name
- chibby family member 1, beta catenin antagonist
- Gene Aliases
- C22orf2, CBY, HS508I15A, PGEA1, PIGEA-14, PIGEA14, arb1
- Chromosome Location
- Chr. 22: 38656653 - 38673850 on Build GRCh38
- UniGene ID
- Hs.334911
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC008839.2 |
- |
2 |
911 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AW613413.1 |
- |
|
336 |
|
Chr. - - [-] |
View Full Product Details