Gene Transcripts

Gene Symbol
DYRK2
Entrez Gene ID
8445
Gene Name
dual specificity tyrosine phosphorylation regulated kinase 2
Gene Aliases
Chromosome Location
Chr. 12: 67648732 - 67665405 on Build GRCh38
UniGene ID
Hs.173135
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_017020032.1 XP_016875521.1 2 7721
GenBank mRNA AK024870.1 - 1 1609
AL832817.1 - 1 3136

View Full Product Details