Gene Transcripts
- Gene Symbol
- CCDC82
- Entrez Gene ID
- 79780
- Gene Name
- coiled-coil domain containing 82
- Gene Aliases
- HSPC048
- Chromosome Location
- Chr. 11: 96352765 - 96389940 on Build GRCh38
- UniGene ID
- Hs.525088
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AF150137.1 |
- |
1 |
680 |
|
Chr. 11 - 96389159 - 96389865 [-] |
GenBankmRNA |
DA249227.1 |
- |
1 |
248 |
|
Chr. 11 - 96388833 - 96389434 [-] |
View Full Product Details