Gene Transcripts
- Gene Symbol
- PWP2
- Entrez Gene ID
- 5822
- Gene Name
- PWP2 periodic tryptophan protein homolog (yeast)
- Gene Aliases
- EHOC-17, PWP2H, UTP1
- Chromosome Location
- Chr. 21: 44107262 - 44131181 on Build GRCh38
- UniGene ID
- Hs.449076
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI968015.1 |
- |
1 |
39 |
|
BU683943.1 |
- |
1 |
98 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK096268.1 |
- |
1 |
2361 |
|
Chr. 21 - 5128592 - 5130994 [+] |
View Full Product Details