Gene Transcripts
- Gene Symbol
- DNAJB14
- Entrez Gene ID
- 79982
- Gene Name
- DnaJ heat shock protein family (Hsp40) member B14
- Gene Aliases
- EGNR9427, PRO34683
- Chromosome Location
- Chr. 4: 99896248 - 99946726 on Build GRCh38
- UniGene ID
- Hs.577426
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA830176.1 |
- |
1 |
131 |
|
Chr. 4 - 99897555 - 99898315 [-] |
GenBankmRNA |
DA307038.1 |
- |
1 |
222 |
|
Chr. 4 - 99897828 - 99898406 [-] |
GenBankmRNA |
DA169403.1 |
- |
1 |
195 |
|
Chr. 4 - 99897803 - 99898379 [-] |
GenBankmRNA |
DA134891.1 |
- |
1 |
13 |
|
Chr. 4 - 99897638 - 99898197 [-] |
View Full Product Details