Gene Transcripts
- Gene Symbol
- ZBTB20
- Entrez Gene ID
- 26137
- Gene Name
- zinc finger and BTB domain containing 20
- Gene Aliases
- DPZF, HOF, ODA-8S, PRIMS, ZNF288
- Chromosome Location
- Chr. 3: 114314500 - 115147280 on Build GRCh38
- UniGene ID
- Hs.202577
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK023302.1 |
- |
1 |
1429 |
|
View Full Product Details