Gene Transcripts
- Gene Symbol
- STK32C
- Entrez Gene ID
- 282974
- Gene Name
- serine/threonine kinase 32C
- Gene Aliases
- PKE, YANK3
- Chromosome Location
- Chr. 10: 132207482 - 132332559 on Build GRCh38
- UniGene ID
- Hs.469002
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BM720428.1 |
- |
1 |
308 |
|
Chr. 10 - 132330291 - 132330659 [-] |
GenBankmRNA |
DN997246.1 |
- |
2 |
542 |
|
Chr. 10 - 132330294 - 132331837 [-] |
GenBankmRNA |
DW426014.1 |
- |
|
171 |
|
Chr. - - [-] |
View Full Product Details