Gene Transcripts

Gene Symbol
STK32C
Entrez Gene ID
282974
Gene Name
serine/threonine kinase 32C
Gene Aliases
PKE, YANK3
Chromosome Location
Chr. 10: 132207482 - 132332559 on Build GRCh38
UniGene ID
Hs.469002
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318878.1 NP_001305807.1 1 136
NM_001318880.1 NP_001305809.1 1 136
NM_001318881.1 NP_001305810.1 1 136
NM_001318882.1 NP_001305811.1 1 136
NM_001318883.1 NP_001305812.1 1 136
XM_011539688.1 XP_011537990.1 1 154
XM_017016100.1 XP_016871589.1 1 154
XM_017016101.1 XP_016871590.1 1 154
GenBank mRNA AK098366.1 - 1 110
AK302102.1 - 1 83
AK308956.1 - 1 114
BC042566.1 - 1 113
CD674243.1 - 1 130
DA574949.1 - 1 113
HY020713.1 - 1 136
HY106710.1 - 1 111


Non Gene Transcripts

Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID Transcript Location
GenBankmRNA DN997246.1 - 1 105 Chr. 10 - 132330294 - 132331837 [-]

View Full Product Details