Gene Transcripts
- Gene Symbol
- NEMF
- Entrez Gene ID
- 9147
- Gene Name
- nuclear export mediator factor
- Gene Aliases
- NY-CO-1, SDCCAG1
- Chromosome Location
- Chr. 14: 49782083 - 49853073 on Build GRCh38
- UniGene ID
- Hs.655964
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AL547508.3 |
- |
1 |
227 |
|
Chr. 14 - 49782094 - 49783001 [-] |
GenBankmRNA |
AL547576.3 |
- |
1 |
227 |
|
Chr. 14 - 49782084 - 49783001 [-] |
GenBankmRNA |
CR622792.1 |
- |
1 |
227 |
|
Chr. 14 - 49782083 - 49783001 [-] |
View Full Product Details