Gene Transcripts
- Gene Symbol
- ZFP92
- Entrez Gene ID
- 139735
- Gene Name
- ZFP92 zinc finger protein
- Gene Aliases
- ZNF897
- Chromosome Location
- Chr. X: 153411461 - 153426481 on Build GRCh38
- UniGene ID
- Hs.534554
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AA450161.1 |
- |
1 |
77 |
|
Chr. X - 153423877 - 153424320 [+] |
GenBankmRNA |
CB150743.1 |
- |
1 |
208 |
|
Chr. X - 153423747 - 153424385 [+] |
View Full Product Details