Gene Transcripts
- Gene Symbol
- C1orf61
- Entrez Gene ID
- 10485
- Gene Name
- chromosome 1 open reading frame 61
- Gene Aliases
- CROC4
- Chromosome Location
- Chr. 1: 156404252 - 156456649 on Build GRCh38
- UniGene ID
- Hs.380027
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095622.1 |
- |
1 |
2358 |
|
View Full Product Details