Gene Transcripts
- Gene Symbol
- ZBTB20
- Entrez Gene ID
- 26137
- Gene Name
- zinc finger and BTB domain containing 20
- Gene Aliases
- DPZF, HOF, ODA-8S, PRIMS, ZNF288
- Chromosome Location
- Chr. 3: 114314500 - 115147280 on Build GRCh38
- UniGene ID
- Hs.202577
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
DA885050.1 |
- |
1 |
126 |
|
Chr. 3 - 114316563 - 114317110 [-] |
GenBankmRNA |
CB957861.1 |
- |
1 |
107 |
|
Chr. 3 - 114316371 - 114317078 [-] |
GenBankmRNA |
BG913239.1 |
- |
|
11 |
|
Chr. 3 - 114316340 - 114316995 [-] |
View Full Product Details