Gene Transcripts
- Gene Symbol
- RAB11B
- Entrez Gene ID
- 9230
- Gene Name
- RAB11B, member RAS oncogene family
- Gene Aliases
- H-YPT3
- Chromosome Location
- Chr. 19: 8390321 - 8404434 on Build GRCh38
- UniGene ID
- Hs.558403
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX647356.1 |
- |
|
1876 |
|
View Full Product Details