Gene Transcripts
- Gene Symbol
- TMEM185B
- Entrez Gene ID
- 79134
- Gene Name
- transmembrane protein 185B
- Gene Aliases
- FAM11B
- Chromosome Location
- Chr. 2: 120217471 - 120223408 on Build GRCh38
- UniGene ID
- Hs.376722
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BI257465.1 |
- |
1 |
413 |
|
Chr. 2 - 120219167 - 120219858 [-] |
GenBankmRNA |
AL119878.1 |
- |
1 |
476 |
|
Chr. 2 - 120219411 - 120219921 [-] |
GenBankmRNA |
DA435851.1 |
- |
1 |
513 |
|
Chr. 2 - 120219391 - 120219956 [-] |
View Full Product Details