Gene Transcripts
- Gene Symbol
- NFYC
- Entrez Gene ID
- 4802
- Gene Name
- nuclear transcription factor Y subunit gamma
- Gene Aliases
- CBF-C, CBFC, H1TF2A, HAP5, HSM, NF-YC
- Chromosome Location
- Chr. 1: 40691570 - 40771603 on Build GRCh38
- UniGene ID
- Hs.713051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK301215.1 |
- |
5 |
902 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK055329.1 |
- |
1 |
1578 |
|
Chr. 1 - 40765293 - 40767975 [+] |
View Full Product Details