Gene Transcripts
- Gene Symbol
- PSPN
- Entrez Gene ID
- 5623
- Gene Name
- persephin
- Gene Aliases
- PSP
- Chromosome Location
- Chr. 19: 6375294 - 6375849 on Build GRCh38
- UniGene ID
- Hs.248159
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK055490.1 |
- |
1 |
310 |
|
- Gene Symbol
- LOC100130856
- Entrez Gene ID
- 100130856
- Gene Name
- uncharacterized LOC100130856
- Gene Aliases
- Chromosome Location
- Chr. : - on Build
- UniGene ID
- Hs.634471
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK095727.1 |
- |
1 |
2042 |
|
View Full Product Details