Gene Transcripts
- Gene Symbol
- YAE1D1
- Entrez Gene ID
- 57002
- Gene Name
- Yae1 domain containing 1
- Gene Aliases
- C7orf36, GK003
- Chromosome Location
- Chr. 7: 39566376 - 39612089 on Build GRCh38
- UniGene ID
- Hs.72168
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648555.1 |
- |
1 |
2763 |
|
View Full Product Details