Gene Transcripts
- Gene Symbol
- ITSN1
- Entrez Gene ID
- 6453
- Gene Name
- intersectin 1
- Gene Aliases
- ITSN, SH3D1A, SH3P17
- Chromosome Location
- Chr. 21: 33642400 - 33899861 on Build GRCh38
- UniGene ID
- Hs.160324
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BX648852.1 |
- |
|
3086 |
|
- Gene Symbol
- MED13L
- Entrez Gene ID
- 23389
- Gene Name
- mediator complex subunit 13 like
- Gene Aliases
- MRFACD, PROSIT240, THRAP2, TRAP240L
- Chromosome Location
- Chr. 12: 115958576 - 116277219 on Build GRCh38
- UniGene ID
- Hs.603766
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK025280.1 |
- |
|
2354 |
|
- Gene Symbol
- MTND1P23
- Entrez Gene ID
- 100887749
- Gene Name
- mitochondrially encoded NADH:ubiquinone oxidoreductase core subunit 1 pseudogene 23
- Gene Aliases
- Chromosome Location
- Chr. 1: 629062 - 629440 on Build GRCh38
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC009345.1 |
- |
|
2529 |
|
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
AK124495.1 |
- |
|
2407 |
|
Chr. - - [-] |
GenBankmRNA |
AK074482.1 |
- |
1 |
670 |
|
Chr. MT - 3307 - 5511 [+] |
View Full Product Details