Gene Transcripts
- Gene Symbol
- FCN1
- Entrez Gene ID
- 2219
- Gene Name
- ficolin 1
- Gene Aliases
- FCNM
- Chromosome Location
- Chr. 9: 134909585 - 134917960 on Build GRCh38
- UniGene ID
- Hs.440898
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AL833097.1 |
- |
1 |
263 |
|
View Full Product Details