Gene Transcripts

Gene Symbol
NFX1
Entrez Gene ID
4799
Gene Name
nuclear transcription factor, X-box binding 1
Gene Aliases
NFX2, TEG-42, Tex42
Chromosome Location
Chr. 9: 33290417 - 33371163 on Build GRCh38
UniGene ID
Hs.413074
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK055847.1 - 1 213

View Full Product Details