Gene Transcripts
- Gene Symbol
- NUDT12
- Entrez Gene ID
- 83594
- Gene Name
- nudix hydrolase 12
- Gene Aliases
- Chromosome Location
- Chr. 5: 103548855 - 103562805 on Build GRCh38
- UniGene ID
- Hs.434289
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC042967.1 |
- |
1 |
597 |
|
View Full Product Details