Gene Transcripts
- Gene Symbol
- MTHFS
- Entrez Gene ID
- 10588
- Gene Name
- 5,10-methenyltetrahydrofolate synthetase (5-formyltetrahydrofolate cyclo-ligase)
- Gene Aliases
- HsT19268
- Chromosome Location
- Chr. 15: 79843547 - 79897285 on Build GRCh38
- UniGene ID
- Hs.194294
- Species
- Homo sapiens
- Gene Symbol
- ST20-MTHFS
- Entrez Gene ID
- 100528021
- Gene Name
- ST20-MTHFS readthrough
- Gene Aliases
- Chromosome Location
- Chr. 15: 79843547 - 79923106 on Build GRCh38
- UniGene ID
- Hs.459049
- Species
- Homo sapiens
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
Transcript Location |
GenBankmRNA |
BX100068.1 |
- |
1 |
272 |
|
Chr. 15 - 79843547 - 79844304 [-] |
View Full Product Details