Gene Transcripts
- Gene Symbol
- TMEM80
- Entrez Gene ID
- 283232
- Gene Name
- transmembrane protein 80
- Gene Aliases
- Chromosome Location
- Chr. 11: 695386 - 705028 on Build GRCh38
- UniGene ID
- Hs.448664
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC038383.1 |
- |
1 |
598 |
|
View Full Product Details